Ethyl-3-chloroimidazo[1,2-a]pyridine-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR01958-1G OR01958-250MG
76561-360EA 92.42 USD
76561-360 76560-226
Ethyl-3-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-3-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09842616
CAS Number: 1000017-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR