1-Fluoronaphthalene-2-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC

PC53519-100MG PC53519-1G PC53519-250MG
76561-194EA 151.78 USD
76561-194 76561-968 76560-194
1-Fluoronaphthalene-2-carboxylic acid 95%
1-Fluoronaphthalene-2-carboxylic acid
Formula: C₁₁H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD16990867
CAS Number: 574-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR