3-Fluoro-4-(methylthio)benzamide 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC3829-1G PC3829-250MG
76560-176EA 579.72 USD
76560-176 76562-464
3-Fluoro-4-(methylthio)benzamide 97%
3-Fluoro-4-(methylthio)benzamide
Formula: C₈H₈FNOS
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00673179
CAS Number: 238742-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR