Potassium carbonate, anhydrous 99%

Supplier: APOLLO SCIENTIFIC

IN14572A-100G IN14572A-1KG
76560-152EA 20.89 USD
76560-152 76563-020
Potassium carbonate, anhydrous 99%
Potassium carbonate
Formula: K₂CO₃
Melting Pt: 891 °C
Density: 2.43 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR