Methyl-2-fluoro-4-nitrobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 2-fluoro-4-nitrobenzoate

PC8742-10G PC8742-1G PC8742-5G
76561-348EA 33.48 USD
76561-348 76559-986 76559-804
Methyl-2-fluoro-4-nitrobenzoate
Methyl 2-fluoro-4-nitrobenzoate
Formula: C₈H₆FNO₄
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD08444028
CAS Number: 392-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR