Imidazo[2,1-b][1,3]benzothiazole-2-carboxylic acid hydrate

Supplier: APOLLO SCIENTIFIC
Warning

OR0880-10G OR0880-1G OR0880-5G
76559-894EA 1732.06 USD
76559-894 76562-096 76561-116
Imidazo[2,1-b][1,3]benzothiazole-2-carboxylic acid hydrate
Imidazo[2,1-b][1,3]benzothiazole-2-carboxylic acid hydrate
Formula: C₁₀H₆N₂O₂S·H₂O
Melting Pt: 266…268 °C
Storage Temperature: Ambient
MDL Number: MFCD08444056
CAS Number: 1284226-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR