Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR01979-1G
OR01979-250MG
76562-684EA
288.54
USD
76562-684
76559-810
Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Formula:
C₁₀H₈Cl₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09842623 CAS Number: 1000018-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...