5-Fluoro-2-oxo-2H-chromene-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC32789-250MG PC32789-1G
76562-928EA 37.7 USD
76562-928 76559-746
5-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
5-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
Formula: C₁₀H₅FO₄
Storage Temperature: Ambient
MDL Number: MFCD26684685
CAS Number: 1540476-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR