Hydrochlorothiazide

Supplier: APOLLO SCIENTIFIC
Danger

OR0507-100G OR0507-25G
76560-006EA 199.28 USD
76560-006 76559-738 76560-700
Hydrochlorothiazide
Hydrochlorothiazide
Formula: C₇H₈ClN₃O₄S₂
Melting Pt: 273…275 °C
MDL Number: MFCD00051765
CAS Number: 58-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR