2-(Dimethylamino)-2-oxoacetic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR0185-1G OR0185-5G
76559-598EA 20.77 USD
76559-598 76562-100
2-(Dimethylamino)-2-oxoacetic acid
2-(Dimethylamino)-2-oxoacetic acid
Formula: C₄H₇NO₃
Melting Pt: 126…128 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01971023
CAS Number: 32833-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR