Tetrahydropyran-4-methanol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR10585-5G
76560-928EA 13.39 USD
76560-928 76560-900 76559-556
Tetrahydropyran-4-methanol 98%
Tetrahydropyran-4-methanol
Formula: C₆H₁₂O₂
Boiling Pt: 105…110 °C/20mmHg
Flash Pt: >61 °C
Storage Temperature: Ambient
MDL Number: MFCD00457804
CAS Number: 14774-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR