3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR019785-1G OR019785-5G
76562-892EA 20.34 USD
76562-892 76559-524
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 98%
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde
Formula: C7H9NO
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00111522
CAS Number: 2199-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR