4-Carboxyphenylboronic acid pinacol ester 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

OR10406-100G OR10406-25G OR10406-5G
76562-720EA 113.08 USD
76562-720 76559-538 76559-462
4-Carboxyphenylboronic acid pinacol ester 98%
4-Carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BO₄
Melting Pt: 227…231 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863710
CAS Number: 180516-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR