CORIO™ CP Refrigerated/Heating Circulators
Supplier: JULABO
9013701.02 9013701.33 9013702.33 9013703.02 9013703.13 9013704.02 9013704.33 9013705.02 9013705.33 9013707.02 9013707.33 9013708.33 9013713.N1.02 9013713.N1.33 9013713.S1.02 9013713.S1.33 9013714.N1.02 9013714.N1.33 9013714.S1.02 9013714.S1.33 9013727.33 9013727.02 9013715.N1.02 9013715.N1.33 9013717.N1.02 9013717.N1.33 9013728.N1.02 9013728.N1.33 9013701.N1.02 9013701.N1.33
76353-836EA 9504.83 USD
76353-836 76353-838 76353-840 76353-842 76353-844 76353-846 76353-848 76353-850 76353-852 76353-854 76353-856 76353-858 76353-860 76353-862 76353-864 76558-220 76558-246 76558-238 76558-168 76558-218 76558-236 76558-254 76558-180 76558-198 76558-160 77442-080 77442-082 77442-084 77442-086 77442-088 77442-090 77442-076 77442-078
CORIO™ CP Refrigerated/Heating Circulators
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
CORIO™ CP refrigerated/heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from –50 to +200 °C.
  • Infinitely adjustable pressure pump
  • Easy to switch between internal and external circulation
  • Bright LED display
  • USB and RS232 ports
For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.
CORIO CP circulators feature a bright LED display button controls. A lever below the display enables simple adjustment between internal and external fluid flow. The USB and RS232 ports facilitate external communication to PC or PLC systems. Pump connections: M16×1 male threads.
To adapt to your individual application there is comprehensive range of accessories.
Certifications: CE, UKCA and DIN 12876-1 Class III.
Ordering information: Each unit is supplied with a pair of 8 and 12 mm hose barbs. Fluid, adapters, tubing sold separately.
Delivery information: Includes CORIO CP immersion circulator, refrigeration unit, mounting bridge, bath tank lid, pair of 8 and 12 mm hose barbs, integrated drain tap.