DYNEO™ DD Refrigerated/Heating Circulators
Supplier: JULABO
9021701.02 9021701.33 9021701.02.D 9021701.33.D 9021701.02.A 9021701.33.A 9021703.02 9021703.13 9021703.02.D 9021703.13.D 9021703.02.A 9021703.13.A 9021704.02 9021704.33 9021704.02.D 9021704.33.D 9021704.02.A 9021704.33.A 9021707.02.D 9021707.33.D 9021707.02.A 9021707.33.A 9021707.02 9021707.33 9021713.N1.02 9021713.N1.02.A 9021713.N1.02.D 9021713.N1.33 9021713.N1.33.A 9021713.N1.33.D 9021713.S1.02 9021713.S1.02.A 9021713.S1.02.D 9021713.S1.33 9021713.S1.33.A 9021713.S1.33.D 9021714.N1.02 9021714.N1.02.A 9021714.N1.02.D 9021714.N1.33 9021714.N1.33.A 9021714.N1.33.D 9021714.S1.02 9021714.S1.02.A 9021714.S1.02.D 9021714.S1.33 9021714.S1.33.A 9021714.S1.33.D 9021727.02 9021727.02.A 9021727.02.D 9021727.33 9021727.33.A 9021727.33.D 9021701.N1.33.A 9021701.N1.33.D 9021715.N1.02 9021715.N1.02.A 9021715.N1.02.D 9021715.N1.33 9021715.N1.33.A 9021715.N1.33.D 9021717.N1.02 9021717.N1.02.A 9021717.N1.02.D 9021717.N1.33 9021717.N1.33.A 9021717.N1.33.D 9021728.N1.02 9021728.N1.02.A 9021728.N1.02.D 9021728.N1.33 9021728.N1.33.A 9021728.N1.33.D 9021701.N1.02 9021701.N1.02.A 9021701.N1.02.D 9021701.N1.33 9021718.N1.33 9021718.N1.33.A 9021718.N1.33.D 9021719.N1.33.D
76289-282EA 9671.94 USD
76289-282 76289-284 76289-286 76289-288 76289-290 76289-292 76289-294 76289-296 76289-298 76289-300 76289-302 76289-304 76289-306 76289-308 76289-310 76289-312 76289-314 76289-316 76289-318 76289-320 76289-322 76289-324 76289-326 76289-328 76289-330 76289-332 76289-334 76289-336 76289-338 76289-340 76289-342 76289-344 76289-346 76289-348 76289-350 76289-352 76289-354 76289-356 76289-358 76289-374 76289-376 76289-378 76289-380 76289-382 76289-384 76558-242 76558-184 76558-172 76558-250 76558-252 76558-264 76558-230 76558-162 76558-232 76558-260 76558-258 76558-274 76558-214 76558-182 76558-256 76558-266 76558-170 76558-164 76558-176 76558-158 76558-192 76558-224 76558-202 76558-244 76558-208 76558-178 76558-226 76558-248 76558-204 76558-186 77442-100 77442-102 77442-104 77442-106 77442-108 77442-110 77442-112 77442-114 77442-116 77442-118 77442-120 77442-122 77442-124 77442-126 77442-128 77442-130 77442-132 77442-134 77442-136 77442-138 77442-092 77442-094 77442-096 77442-098 77971-800 77971-802 77971-804 77971-806
DYNEO™ DD Refrigerated/Heating Circulators
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
DYNEO™ DD refrigerated/heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from –20 to 200 to –50 to 200 °C.
  • Infinitely adjustable pressure pump
  • Easy switching between internal and external circulation
  • Large color TFT display, multilingual interface
  • Integrated programmer
  • PT100 port facilitates external temperature control via PT100 sensor
  • USB port
  • Optional RS232 interface or analog interfaces
  • Includes basket to hold up to 20 bottles of 0.5 L size
  • Includes clear polycarbonate lid
For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.
DYNEO DD circulator features a color TFT display with central knob controller for operating/programming. Integrated programmer. A lever below the display enables simple adjustment between internal and external fluid flow. The USB port facilitates external communication with models options to add RS232 interface or analog interface. Operation in °C or °F. Pump connections: M16×1 male threads.
Certifications: CE, UKCA. DIN 12876-1 Class III.
Ordering information: Each unit is supplied with a pair of 8 and 12 mm hose barbs. Fluid, adapters, tubing sold separately.
Delivery information: Includes DYNEO DD immersion circulator, refrigeration unit, mounting bridge, bath tank lid, pair of 8 and 12 mm hose barbs, integrated drain tap.