Anti-IL-12 p75 Rat Monoclonal Antibody [clone: R2-9A5]

Supplier: Bio X Cell
BE0233-25MG BE0233-1MG BE0233-5MG BE0233-50MG BE0233-100MG
103253-144EA 3920 USD
103253-144 103253-146 103253-148 103253-150 103572-586 76555-810 76556-384 76556-576 76557-030 76557-224
Anti-IL-12 p75 Rat Monoclonal Antibody [clone: R2-9A5]
Antibodies
InVivoMab anti-mouse IL-12 p75, Clone: R2-9A5, Isotype: rat IgG2b, Applications: in vivo IL-12p75 neutralization, ELISA

Type: Primary
Antigen: IL-12 p75
Clonality: Monoclonal
Clone: R2-9A5
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR