Anti-CD8 alpha Rat Monoclonal Antibody [clone: 2.43]

Supplier: Bio X Cell
BE0061-50MG BE0061-100MG BE0061-25MG BE0061-1MG BE0061-5MG
103253-574EA 3920 USD
103253-574 103253-576 103253-578 103253-580 103572-496 76557-012 76557-178 76557-208 76557-238 76557-302
Anti-CD8 alpha Rat Monoclonal Antibody [clone: 2.43]
Antibodies
InVivoMab anti-mouse CD8α, Clone: 2.43, Isotype: rat IgG2b, κ, Applications: in vivo CD8+ T cell depletion

Type: Primary
Antigen: CD8alpha
Clonality: Monoclonal
Clone: 2.43
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR