Anti-CSF1R (CD115) Rat Monoclonal Antibody [clone: AFS98]

Supplier: Bio X Cell
BE0213-50MG BE0213-1MG BE0213-100MG BE0213-25MG BE0213-5MG
103253-984EA 3920 USD
103253-984 103253-986 103253-988 103253-990 103572-560 76554-498 76555-024 76555-690 76556-132 76556-800
Anti-CSF1R (CD115) Rat Monoclonal Antibody [clone: AFS98]
Antibodies
InVivoMab anti-mouse CSF1R (CD115), Clone: AFS98, Isotype: rat IgG2a, κ, Applications: in vivo macrophage depletion, in vitro CSF-R1 neutralization, in vivo monocyte depletion, Flow cytometry

Type: Primary
Antigen: CSF1R (CD115)
Clonality: Monoclonal
Clone: AFS98
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR