Anti-MHC Class II (I-Ek/RT1-D) Mouse Monoclonal Antibody [clone: 14-4-4S (HB-32)]

Supplier: Bio X Cell
BE0167-1MG BE0167-100MG BE0167-50MG BE0167-25MG BE0167-5MG
103253-864EA 3920 USD
103253-864 103253-866 103253-868 103253-870 103572-856 76554-524 76554-718 76555-908 76555-990 76556-004
Anti-MHC Class II (I-Ek/RT1-D) Mouse Monoclonal Antibody [clone: 14-4-4S (HB-32)]
Antibodies
InVivoMab anti-mouse/rat MHC Class II (I-Ek/RT1-D), Clone: 14-4-4S (HB-32), Isotype: mouse IgG2a, κ, Applications: in vivo blocking of antigen presentation, Flow cytometry

Type: Primary
Antigen: MHC Class II (I-Ek/RT1-D)
Clonality: Monoclonal
Clone: 14-4-4S (HB-32)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR