Anti-CD54 (ICAM-1) Mouse Monoclonal Antibody [clone: R6-5-D6]

Supplier: Bio X Cell
BE0020-2-1MG BE0020-2-25MG BE0020-2-100MG BE0020-2-50MG BE0020-2-5MG
103252-342EA 3920 USD
103252-342 103252-344 103252-346 103252-348 103572-440 76554-570 76555-734 76555-980 76556-300 76557-340
Anti-CD54 (ICAM-1) Mouse Monoclonal Antibody [clone: R6-5-D6]
Antibodies
InVivoMab anti-human CD54 (ICAM-1), Clone: R6-5-D6, Isotype: mouse IgG2a, Applications: in vitro T cell stimulation/activation, Immunofluorescence

  • Recommended Dilution Buffer: IP0065

Type: Primary
Antigen: CD54 (ICAM-1)
Clonality: Monoclonal
Clone: R6-5-D6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR