Anti-Ly6G/Ly6C Rat Monoclonal Antibody [clone: NIMP-R14]

Supplier: Bio X Cell
BE0320-50MG BE0320-25MG BE0320-1MG BE0320-100MG BE0320-5MG
103778-418EA 3920 USD
103778-418 103778-420 103778-422 103778-424 103778-426 76554-670 76555-482 76555-522 76555-958 76556-478
Anti-Ly6G/Ly6C Rat Monoclonal Antibody [clone: NIMP-R14]
Antibodies
Type: Primary
Antigen: Ly6G/Ly6C
Clonality: Monoclonal
Clone: NIMP-R14
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR