Anti-IFNAR-1 Mouse Monoclonal Antibody [clone: MAR1-5A3]

Supplier: Bio X Cell
BE0241-25MG BE0241-50MG BE0241-1MG BE0241-5MG BE0241-100MG
103254-080EA 3920 USD
103254-080 103254-082 103254-084 103254-086 103572-602 76554-634 76554-648 76555-112 76555-174 76556-514
Anti-IFNAR-1 Mouse Monoclonal Antibody [clone: MAR1-5A3]
Antibodies
InVivoMab anti-mouse IFNAR-1, Clone: MAR1-5A3, Isotype: mouse IgG1, κ, Applications: in vivo IFNAR-1 blockade, in vitro IFNAR-1 blockade

Type: Primary
Antigen: IFNAR-1
Clonality: Monoclonal
Clone: MAR1-5A3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR