Anti-CD28 Mouse Monoclonal Antibody [clone: JJ319]

Supplier: Bio X Cell
BE0040-50MG BE0040-5MG BE0040-100MG BE0040-1MG BE0040-25MG
103252-478EA 3920 USD
103252-478 103252-480 103252-482 103252-484 103572-476 76554-620 76555-104 76555-946 76556-880 76556-952
Anti-CD28 Mouse Monoclonal Antibody [clone: JJ319]
Antibodies
InVivoMab anti-rat CD28, Clone: JJ319, Isotype: mouse IgG1, κ, Applications: in vitro T cell stimulation/activation, Flow cytometry

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: JJ319
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR