Anti-CD40L (CD154) Armenian Hamster Monoclonal Antibody [clone: MR-1]

Supplier: Bio X Cell
BE0017-1-1MG BE0017-1-5MG BE0017-1-25MG BE0017-1-100MG BE0017-1-50MG
103252-310EA 3920 USD
103252-310 103252-312 103252-314 103252-316 103572-432 76554-962 76556-156 76557-054 76557-094 76557-172
Anti-CD40L (CD154) Armenian Hamster Monoclonal Antibody [clone: MR-1]
Antibodies
InVivoMab anti-mouse CD154 (CD40L), Clone: MR-1, Isotype: Armenian hamster IgG, Applications: in vivo blocking of CD40/CD40L signaling, in vitro blocking of CD40/CD40L signaling

  • Recommended Dilution Buffer: IP0070

Type: Primary
Antigen: CD40L (CD154)
Clonality: Monoclonal
Clone: MR-1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR