Anti-KLH Rat Monoclonal Antibody [clone: LTF-2]

Supplier: Bio X Cell
BE0090-5MG BE0090-1MG BE0090-25MG BE0090-100MG BE0090-50MG
103252-582EA 3920 USD
103252-582 103252-584 103252-586 103252-588 103572-788 76554-454 76554-610 76554-910 76555-162 76557-066
Anti-KLH Rat Monoclonal Antibody [clone: LTF-2]
Antibodies
The LTF-2 monoclonal antibody reacts with keyhole limpet hemocyanin (KLH). Because KLH is not expressed by mammals this antibody is ideal for use as an isotype-matched control for rat IgG2b antibodies in most in vivo and in vitro applications.

Type: Control
Antigen: KLH
Clonality: Monoclonal
Clone: LTF-2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR