Klercide™ mop wipe frame, stainless steel
Supplier: ECOLAB LIFE SCIENCES
3036310
76554-276EA 615.61 USD
76554-276
Klercide™ mop wipe frame, stainless steel
Mops
This stainless steel mop frame features a stiff movable flat top.
  • Fully autoclavable
  • Made from stainless steel with stiff, moveable flat mop head frame
  • Designed for use with Klercide™ pre-saturated mop wipes
  • Supplied Gamma irradiated
Option of adding Klercide™ silicone pad for better contact on uneven surfaces.