Klerwipe™ 70/30 IPA mop wipes
Supplier: ECOLAB LIFE SCIENCES
6600121
76554-270CS 705.86 USD
76554-270
Klerwipe™ 70/30 IPA mop wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Klerwipe™ 70/30 IPA mop wipes are ready-to-use mop wipes for cleaning and residue removal on large surfaces. Sterile non-woven, low particulate wipes impregnated with 70% v/v Isopropyl alcohol blended with Water for Injection.
  • Made from hydroentangled polyester/cellulose
  • Consistent Cleaning: Pre-saturated with 0.2 micron-filtered 70% isopropyl alcohol solution blended with WFI
  • Ready-to-Use Mop Wipes: Pre-saturated mop wipes remove the need for mop and bucket systems, the ready to use feature is ideal for streamlining compliant cleaning protocols for larger surfaces in the cleanroom
  • Premium Grade Substrate: High-quality polyester-cellulose mop wipes are low-linting and pre-laundered in an ISO Class 4 cleanroom to significantly reduce particles
  • Sterile Production: Ultrasonically sealed and gamma irradiated in a Grade C/ISO Class 5 cleanroom; supported with certificates of analysis and Certificates of Sterility with every batch
  • Convenient Packaging: Ready-to-use mop wipes are double bagged to streamline movement into high-grade areas, with an easily peelable pack for easy opening with gloved hands
For cleaning large cleanroom surfaces, Klerwipe™ 70/30 Mop Wipes are a quick and convenient method.
Streamline daily cleaning and residue removal protocols for walls, floors and other large surfaces, with the simplicity of ready-to-use sterile mop wipes pre-saturated with 70/30 IPA blended with WFI.
The wipes are pre-saturated with 70% v/v isopropyl alcohol blended with Water for Injection (WFI). Designed to be used with mop frames, the wipes are hydroentangled polyester/cellulose, non-woven and binder free.
Basis weight: 120-140 g/m²