Klerwipe™ low residue Peroxide mop wipes
Supplier: ECOLAB LIFE SCIENCES
6600122
76554-254CS 1175.23 USD
76554-254
Klerwipe™ low residue Peroxide mop wipes
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Klerwipe™ Low Residue Peroxide Mop Wipes are sterile ready-to-use mop wipes for high-grade cleaning with minimal residue. The non-woven, low particulate wipes are made from hydroentangled polyester/cellulose and impregnated with 6% Hydrogen Peroxide.
  • Powerful Cleaning and Residue Removal: Unique formulation of 6% hydrogen peroxide is designed specifically for use in cleanrooms and other controlled areas, providing an effective alternative to IPA in general cleaning protocols
  • Ready-to-Use Mop Wipes: Pre-saturated mop wipes remove the need for mop and bucket systems, the ready to use feature is ideal for streamlining compliant cleaning protocols for larger surfaces in the cleanroom
  • Premium-Grade Substrate: High-quality polyester-cellulose mop wipes remain wet throughout use and are low-linting and pre-laundered in an ISO Class 4 cleanroom to significantly reduce particles
  • Sterile Production: Mop wipes are pre-saturated with 0.2 micron-filtered solution, ultrasonically sealed and gamma irradiated in a Grade C/ISO Class 5 cleanroom; supported with certificates of analysis and Certificates of Sterility with every batch
  • Convenient Packaging: Ready-to-use mop wipes are double bagged to streamline movement into high-grade areas, with an easily peelable pack for easy opening with gloved hands
  • Designed to be easily fitted to Klercide™ mop wipe frames
For removing surface contamination on cleanroom surfaces, Klerwipe™ Low Residue Peroxide Mop Wipes are ideal for routine and rotational use. They are ideal for streamline cleaning and residue removal in smaller high-grade areas where a low-residue product is required.
Designed to leave minimal residues, the wipes are presaturated with 6% hydrogen peroxide and Water for Injection (WFI) and filled, sealed, packed and bagged in an ISO Class 5 cleanroom.
Basis weight: 58-78 g/m²