Klercide™ Low Residue Peroxide
Supplier: ECOLAB LIFE SCIENCES
6000187
76554-252CS 695.02 USD
76554-252
Klercide™ Low Residue Peroxide
Cleaning Agents Disinfectants
Klercide™ low residue peroxide is a sterile hydrogen peroxide solution with low residues designed for pharmaceutical cleanrooms.
  • Powerful cleaning and residue removal / designed specifically for use in cleanrooms and other controlled areas where a low-residue product is required, providing an effective alternative to IPA in general cleaning protocols.
  • Low residue, no-rinse use - low-residue formulation enables highly effective cleaning and reside removal / without the need to wipe down with alcohol or water afterwards / streamlining cleaning protocols.
  • Ready-to-use formats - available in trigger spray and capped bottle formats / all designed to reduce handling and need for dilution, to streamline cleaning protocols and reduce cost-to-clean.
  • Sterile delivery system - trigger-spray format features patented SteriShield™ delivery system (SDS), providing sterility assurance of contents in bottle by forming a complete seal that prevents any air from (microbial or chemical contaminants) being drawn back into the bottle.
  • Cleanroom-grade sterile production - solution is 0.2 micron-filtered and filled under unidirectional Grade A/ISO Class 7 airflow in a Grade B/ISO Class 5 cleanroom / and double-bagged to simplify movement into high-grade areas. Supported with Certificates of analysis and sterility with every batch.
For use in cleaning and removing residues, Klercide™ Low Residue Peroxide is suitable for use on all types of hard surfaces in cleanrooms, controlled environments, and isolators. The product is a blend of high grade 6% hydrogen peroxide and Water For Injection (WFI).
Klercide low residue peroxide is a 0.2 micron filtered, bagged and terminally sterilized by gamma irradiation. Endotoxin tested to an approved pharmacopeia method.