Klercide™ triple bucket system
Supplier: ECOLAB LIFE SCIENCES
3038200
76554-226EA 4545.86 USD
76554-226
Klercide™ triple bucket system
Mops
Ecolab’s Klercide™ Triple Bucket System features a unique controlled application wringer which enables the even distribution of disinfectant over cleanroom surfaces. This enables accurate, repeatable results which help minimize operator error and enhance compliance with cGMP.
  • Includes “click wringer” for controlled application that helps prevent potential fluid application variations between operators and ensures the optimum cleaning and disinfection results every time
  • Includes 3 large buckets for segregation of solutions and waste, 4 small buckets for the top of the trolley and Klercide stainless steel flat mop frame and handle
  • Fully autoclavable