Klercide™ Isolator Cleaning Tool
Supplier: ECOLAB LIFE SCIENCES
92023002
76554-220EA 618.95 USD
76554-220
Klercide™ Isolator Cleaning Tool
Mops
The Klercide™ Isolator Cleaning Tool and Cleaning Pads are specifically designed for isolator, laminar flow cabinet cleaning and other cleanroom areas where access is restricted.
  • Fully autoclavable
  • Lightweight for ease of use
  • Made from 316 stainless steel with 600 mm reach
  • Supplied cleanroom processed, double bagged and sterilized by gamma irradiation
  • Compatible with the Klercide™ range of alcohols and biocides
The Klercide™ Isolator and Laminar Flow Cabinet Cleaning Tool consists of a two-part 316 stainless steel handle and a specially shaped head.
Designed for use with sterile single use cleaning pads.