COSA™ Oxonia Active Agent
Supplier: ECOLAB LIFE SCIENCES
6600027
76554-210EA 5491.13 USD
76554-210
COSA™ Oxonia Active Agent
Cleaning Agents Detergents and Surface Cleansers
COSA™ Oxonia Active is a peroxyacetic acid antimicrobial agent.
  • Promotes Quality Assurance
  • Effective against many pathogenic organisms, including spore-forming microorganisms.
  • COSA™ Oxonia Active is supported by a complete validation support package including methods for residue quantitation.
  • Non-corrosive to 304 or 316 stainless steel and aluminum surfaces when used at recommended concentrations.
  • Non-foaming formulation minimizes CIP cycle time and improves efficiency.
  • Compatible with most plastic and rubber materials.
Ecolab COSA™ Oxonia Active is an EPA-registered, peroxyacetic acid, antimicrobial agent with broad antimicrobial activity for use in a number of sanitizing applications in the life science industry, as well as for sterilization of aseptic processing and packaging equipment.
EPA Registered disinfectant and sanitizer.
EPA Reg #1677-129