COSA™ CIP 92 Alkaline Cleaner
Supplier: ECOLAB LIFE SCIENCES
6300542
76554-198EA 607.91 USD
76554-198 76554-200
COSA™ CIP 92 Alkaline Cleaner
Cleaning Agents Detergents and Surface Cleansers
COSA™ CIP 92 is a liquid, alkaline cleaner.
  • Promotes Quality Assurance
  • Excellent removal of organic soils such as fat, oil, oil-in-water emulsions, and water-in-oil emulsions.
  • Suitable for cleaning Carbopol® and similar polymeric materials.
  • Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation.
COSA™ CIP 92 is a alkaline surfactant-containing cleaning agent for the removal of emulsion- and fat-containing residues in clean in place (CIP) pharmaceutical processing equipment including piping, mixers, fluid beds, tanks, and storage vessels.