COSA™ CIP 90 Alkaline Cleaner
Supplier: ECOLAB LIFE SCIENCES
6301054
76554-194EA 540.37 USD
76554-194 76554-196
COSA™ CIP 90 Alkaline Cleaner
Cleaning Agents Detergents and Surface Cleansers
Cosa™ CIP 90 is a mildly alkaline cleaner.
  • Mild alkaline formula allows for dual use in CIP or manual applications
  • Powerful blend of surfactants and sequestrants for broad cleaning ability
  • Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation
For use in CIP pharmaceutical and cosmetic processing equipment, COSA™ CIP 90 is a mildly alkaline, surfactant containing cleaner. The product has broad cleaning ability and is ideal for the removal of organic soils such as fat, oil, and oil-in-water emulsions in piping, mixers, fluid beds, tanks, and storage vessels. The near-neutral formula also allows for manual cleaning.