Hydraulic Ram Discharge System, MorehouseCowles
Supplier: MorehouseCowles
491653
76553-868EA 50397.7 USD
76553-868
Hydraulic Ram Discharge System, MorehouseCowles
Homogenizers Benchtop Homogenizers
Your heavy, viscous products can be effortlessly removed from the tank when you activate the MorehouseCowles hydraulic Ram discharge system. With a powerful push, the product is forced out of the dispersion tank, so it takes less labor and time to empty and clean the equipment.
  • Avoids product waste
Delivery information: Comes completely assembled and ready to use.
Packaging: Shipped in a wooden crate.