Compact Shakers, Yamato
Supplier: Yamato Scientific
MK-161-115V
76550-266EA 3204.15 USD
76550-266 76550-270
Compact Shakers, Yamato
Shakers and Mixers Rotating/Rocking Shakers
The compact design of this shaker saves precious space.
  • Rotary shaking and reciprocate range: 30 mm
  • Various shaking modes included
  • Shaking frequency: 20 to ~200 rpm, random adjustment
  • Digital display of shaking frequency
  • Equipped with shaking timer
It features changeable rotary and reciprocate motions to mix, extract, and stir samples.
A shaking timer is provided, and a digital display indicates shaking frequency. The unit works with IN602CSW incubator for shaking incubation.
Accessories are easy to add and remove.