Magnetic Stirrers with Hot Plate, Yamato
Supplier: Yamato Scientific
MG-600H-115V
76550-250EA 4212.47 USD
76550-250 76550-254 76550-258 76550-262
Magnetic Stirrers with Hot Plate, Yamato
Stirrers Magnetic Hotplate Stirrers
These magnetic stirrers include ceramic-coated aluminum plates.
  • Stirring rate up to 1500 rpm
  • Stirring capacity up to 10000 ml
  • Chemical-Resistant plate
  • Triac input temperature control
The plates are chemical resistant and heat conductive.
Each magnetic stirrer includes a stirrer bar.