Benchtop Water Circulator, CB100, Yamato
Supplier: Yamato Scientific
CB-100-115V
76550-228EA 3643.36 USD
76550-228
Benchtop Water Circulator, CB100, Yamato
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Supplies a source of temperature controlled fluid, typically water, which removes heat from a process.
  • Wide temperature range of −10 ~ +80 °C
  • High head and flow rate
  • Easy to clean intake filter
  • Standard equipped with drain
  • Nozzle can be used in any orientation
  • Compact
  • Low GWP value that is friendly to the global environment
Keeps water in the condenser at a stable low temperature thereby creating ideal conditions for collecting the maximum amount of solvent.