Self Laminating Labels
Supplier: CARESFIELD, LLC
83178
76550-096CS 576.03 USD
76550-096
Self Laminating Labels
Labels
Caresfield self-laminating direct-print laser wristbands allow printing wristbands and labels on one sheet with your laser printer.
  • Laser printer required
  • Produce high-quality text and graphics
  • Often used for admitting purpose in healthcare facilities
  • Self-laminating (protects barcodes and printed text)
  • Tamper-evident, adhesive closure
  • Not made with natural rubber latex