Thermal Transfer Printer Label
Supplier: CARESFIELD, LLC
82343
76550-068CS 190.05 USD
76550-068
Thermal Transfer Printer Label
Labels
Direct thermal label roll with multipart labels.
  • Suitable for applications not requiring a long shelf life
  • No ink, toner, or ribbon required
  • Sensor bar included