VWR® Microplate Mixer
Supplier: VWR International
BT1500-VWR
76549-934EA 1404.48 USD
76549-934
VWR® Microplate Mixer
Shakers and Mixers Microplate Shakers
VWR® microplate mixer with platform for up to 4 microplates.
  • Includes platform for up to four microplates
  • Digital speed control up to 1500 rpm
  • Optional adapter for mixing microtubes
  • Provides thorough and uniform mixing in microplates
VWR® vortex mixer with microplate platform provides high speed vortexing (2750 rpm) with touch activation.
Certifications: CE marked