Formalin Neutralizing Pad for Model LD500 - LD511 Tissue Container Containment Trays, Mortech
Supplier: Mortech Manufacturing
LD511
76549-904EA 291.28 USD
76549-904
Formalin Neutralizing Pad for Model LD500 - LD511 Tissue Container Containment Trays, Mortech
Surface Protectors Equipment Protectors
Formalin Neutralizing pads for use in tissue containment trays found in portable shelving units.
  • For use with LD501 tissue container containment trays
  • Five per package