Biopsy Bags, Mortech
Supplier: Mopec
BI001 BI002 BI003 BI004 BI005
76549-766EA 490.08 USD
76549-766 76549-768 76549-770 76549-772 76549-774
Biopsy Bags, Mortech
Bags Sample Bags
Easy to open and available in three convenient sizes, you’ll find our nylon mesh biopsy bags are the optimal solution for protecting specimens during tissue processing.
  • Ensure unrestricted fluid movement around tissue
  • Made from nylon, 0.2 mm mesh dia.
  • All seams are heat-sealed
  • Reduce reagent carryover and pressure artifacts often found with sponges
  • Rapidly remove specimen with convenient peel-open bag
  • Reduces the loss of small biopsy specimens
Available in standard or premium grade.