Mopec Melody Shielding Spray, Mortech
Supplier: MORTECH MANUFACTURING
BG300
76549-744EA 36.89 USD
76549-744
Mopec Melody Shielding Spray, Mortech
Stains Microscopy and staining reagents
Applied after a specimen is marked with a dye, Melody is a quick drying fixative spray that forms a protective layer on the marked surface while still allowing the tissue processing reagents to penetrate the tissue. Melody is unique to Mopec and can be used with all dye colors and on all specimen types. The chemistry of pigments, especially those seen in yellow, orange and red tissue marking dyes manufactured by alternative suppliers often suffer during tissue processing protocols.
  • Quick-drying fixative spray
  • Enables strong adhesion of dyes on particularly fatty tissues
  • Ensures marked margins (including yellow, orange and red) are durable and unaffected during tissue processing
  • Can be used with all colors of Harmony dyes and all specimen types
Melody Ink Shielding Spray has been specifically designed for use with our Harmony range and ensures that specimen margins marked with our dyes (including the colors mentioned above) are durable and unaffected during tissue processing.
Reduced Leaching of dyes help reduce the clogging up of the processors, eliminating potential failure and downtime.