Face Shield, Full Length, Mortech
Supplier: Mortech Manufacturing
BE197
76549-684EA 26.13 USD
76549-684
Face Shield, Full Length, Mortech
Face Shields
Reusable, easy to clean, face shields provide lightweight, full face protection and can be comfortably worn with eyeglasses. Constructed from latex free, fiberglass free and distortion free durable plastic. The window features an anti-fog and anti-static coating for all day wear. Foam headband relieves pressure and provides added comfort throughout.
  • Used for everyday protection, prevent fog, prevent droplets and splash
  • Clear
  • Transparent box feature
  • General use, non-surgical
Note: Remove outer plastic protective film before using.