No-Contact Infrared Thermometer, HT-820D, Mortech
Supplier: Mopec
BE195
76549-680EA 123.25 USD
76549-680
No-Contact Infrared Thermometer, HT-820D, Mortech
Thermometers Digital Thermometers
The HT-820D Body Infrared Thermometer is designed to be a screening device to measure body and forehead temperature without contact. The easily visible three-color alarm backlight display makes it easy to detect higher temperatures during temperature measurement.
  • Easily change units from °C to °F in 2 steps
  • Accuracy is ±0.3 °C (±0.6 °F)
  • Internal memory stores up to 34 readings for easy recall
  • Three color alarm backlight display visually show normal, slightly high, and serious temperatures