Face Mask, KN95, Mortech
Supplier: Mopec
BE194
76549-678EA 60.4 USD
76549-678
Face Mask, KN95, Mortech
Masks Hygiene Masks
Professional protection, disposable medical mask.
  • White
  • General use, non-surgical
Nonsterile. Filtration efficiency 95%. Flexible elastic ear tape. Multilayer filtration.
Certifications: FDA and CE approved.