No Contact Infrared Thermometer, JRT200, Mortech
Supplier: Mopec
BE193
76549-676EA 195.68 USD
76549-676
No Contact Infrared Thermometer, JRT200, Mortech
Thermometers Digital Thermometers
Featuring a large screen tri-color backlight, infrared contact measuring forehead temperature. Can store 32 sets of data.
  • 99.9 accurate temperature measurement
  • One second fast and convenient measurement
  • Non-contact ear or forehead measurement
  • Dual mode
  • Infrared sensor
  • Automatically shutdown during waiting
Certifications: FDA and CE certified and approved.