Replacement Cast Vacuum Filter For Stryker 986 Cast Vac, Mortech
Supplier: Mortech Manufacturing
BD986-500
76549-628EA 335.96 USD
76549-628
Replacement Cast Vacuum Filter For Stryker 986 Cast Vac, Mortech
Filters Venting Filters
The BD986-500 vacuum filter is a brand new direct replacement for the Stryker 986-500 Cast Saw Vac filter. It can be used in both the Styker 986 Mobile and 987 Wall Mount vacuums. Replacing the filter every 6 months more more is recommended depending on usage.