Raised Body Boards, Mortech
Supplier: Mortech Manufacturing
BC081
76549-612EA 371.41 USD
76549-612
Raised Body Boards, Mortech
Ergonomic Supports Back Supports
Body boards are used to raise the body above the work surface to allow for proper fluid drainage.
  • Machined from a single piece of high density polyethylene
  • Boards raise the body above the work surface allowing for added fluid drainage
  • Non-skid rubber feet are placed where contact is made with the work surface
  • Set of 3